N-Methyl-4-boronobenzenesulphonamide 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD45847-1GEA
1140
DKK
BLDPBD45847-1G
BLDPBD45847-5G
BLDPBD45847-250MG
N-Methyl-4-boronobenzenesulphonamide 98%
N-Methyl-4-boronobenzenesulphonamide
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₇H₁₀BNO₄S Molekylvægt: 215,03 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD06659864 CAS-nummer: 226396-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...