1,4-Diamino-2,3-dihydroanthraquinone 99%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD4562-100GEA 334 DKK
BLDPBD4562-100G BLDPBD4562-500G BLDPBD4562-25G BLDPBD4562-5G
1,4-Diamino-2,3-dihydroanthraquinone 99%
1,4-Diamino-2,3-dihydroanthraquinone

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₄H₁₂N₂O₂
Molekylvægt: 240,2600 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00035689
CAS-nummer: 81-63-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR