2-Fluoro-N-methylbenzylamine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD44313-25GEA 1200 DKK
BLDPBD44313-25G BLDPBD44313-5G BLDPBD44313-100G BLDPBD44313-1G BLDPBD44313-10G
2-Fluoro-N-methylbenzylamine 98%
2-Fluoro-N-methylbenzylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₀FN
Molekylvægt: 139,17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04629633
CAS-nummer: 399-30-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR