4-Amino-3-chlorobenzonitrile 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD4369-25GEA 264 DKK
BLDPBD4369-25G BLDPBD4369-100G BLDPBD4369-1G BLDPBD4369-10G BLDPBD4369-500G BLDPBD4369-5G
4-Amino-3-chlorobenzonitrile 98%
4-Amino-3-chlorobenzonitrile

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₅ClN₂
Molekylvægt: 152,58 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052917
CAS-nummer: 21803-75-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR