(+)-Taxifolin 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD43158-250MGEA 357 DKK
BLDPBD43158-250MG BLDPBD43158-1G BLDPBD43158-5G BLDPBD43158-100MG
(+)-Taxifolin 98%
(+)-Taxifolin

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₁₅H₁₂O₇
Molekylvægt: 304,2516 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD15146487
CAS-nummer: 480-18-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR