2-Brom-4-(trifluormethoxy)anilin 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD4302-25GEA 125 DKK
BLDPBD4302-25G BLDPBD4302-500G BLDPBD4302-10G BLDPBD4302-5G BLDPBD4302-100G
2-Brom-4-(trifluormethoxy)anilin 98%
2-Brom-4-(trifluormethoxy)anilin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₅BrF₃NO
Molekylvægt: 256,02 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00203478
CAS-nummer: 175278-17-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR