3,4-Difluoro-2-methylbenzonitrile 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD42054-1GEA 202 DKK
BLDPBD42054-1G BLDPBD42054-25G BLDPBD42054-10G BLDPBD42054-5G
3,4-Difluoro-2-methylbenzonitrile 98%
3,4-Difluoro-2-methylbenzonitrile

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₅F₂N
Molekylvægt: 153,13 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07777154
CAS-nummer: 847502-83-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR