Glycin 99%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD41802-25GEA 46.6 DKK
BLDPBD41802-25G BLDPBD41802-100G BLDPBD41802-500G BLDPBD41802-0.025G
Glycin 99%
Glycin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: NH₂CH₂COOH
Molekylvægt: 75,07 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008131
CAS-nummer: 56-40-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR