5-Iodo-2,4-dimethoxypyrimidine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD41760-5GEA 473 DKK
BLDPBD41760-5G BLDPBD41760-1G BLDPBD41760-250MG BLDPBD41760-25G
5-Iodo-2,4-dimethoxypyrimidine 95%
5-Iodo-2,4-dimethoxypyrimidine

Storage condition: Keep in dark place, Sealed in dry, Store in freezer, under -20°C

Formel: C₆H₇IN₂O₂
Molekylvægt: 266,04 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00090865
CAS-nummer: 52522-99-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR