(4-Nitrophenyl)methanesulphonyl chloride 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD41667-5GEA 256 DKK
BLDPBD41667-5G BLDPBD41667-250MG BLDPBD41667-10G BLDPBD41667-25G BLDPBD41667-1G
(4-Nitrophenyl)methanesulphonyl chloride 95%
(4-Nitrophenyl)methanesulphonyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₆ClNO₄S
Molekylvægt: 235,64 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00034030
CAS-nummer: 4025-75-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR