2,3,4-Trifluorotoluene 99%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD41417-1GEA 77.6 DKK
BLDPBD41417-1G BLDPBD41417-5G
2,3,4-Trifluorotoluene 99%
2,3,4-Trifluorotoluene

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₅F₃
Molekylvægt: 146,1098 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00075194
CAS-nummer: 193533-92-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR