2-Chloro-4'-phenylacetophenone 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD4133-1GEA 132 DKK
BLDPBD4133-1G
2-Chloro-4'-phenylacetophenone 98%
2-Chloro-4'-phenylacetophenone

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₁₄H₁₁ClO
Molekylvægt: 230,6895 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00058937
CAS-nummer: 635-84-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR