4-Bromo-2-nitro-6-(trifluoromethyl)aniline 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD41043-5GEA 907 DKK
BLDPBD41043-5G BLDPBD41043-25G BLDPBD41043-1G
4-Bromo-2-nitro-6-(trifluoromethyl)aniline 98%
4-Bromo-2-nitro-6-(trifluoromethyl)aniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₄BrF₃N₂O₂
Molekylvægt: 285,02 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09835318
CAS-nummer: 157026-18-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR