2,6-Dichloro-3-fluorobenzoic acid 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD4068-5GEA 628 DKK
BLDPBD4068-5G BLDPBD4068-1G BLDPBD4068-25G BLDPBD4068-100G
2,6-Dichloro-3-fluorobenzoic acid 98%
2,6-Dichloro-3-fluorobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₃Cl₂FO₂
Molekylvægt: 209,0019 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06658165
CAS-nummer: 178813-78-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR