6-Bromo-2-naphthylamine 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD36939-100GEA
3980
DKK
BLDPBD36939-100G
BLDPBD36939-500G
BLDPBD36939-10G
BLDPBD36939-5G
BLDPBD36939-250MG
BLDPBD36939-25G
BLDPBD36939-1G
6-Bromo-2-naphthylamine 98%
6-Bromo-2-naphthylamine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₁₀H₈BrN Molekylvægt: 222,0812 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01026466 CAS-nummer: 7499-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...