2-Amino-6-methoxy-3-nitropyridine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD3637-5GEA 62.1 DKK
BLDPBD3637-5G BLDPBD3637-100G BLDPBD3637-25G
2-Amino-6-methoxy-3-nitropyridine 97%
2-Amino-6-methoxy-3-nitropyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₇N₃O₃
Molekylvægt: 169,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03206481
CAS-nummer: 73896-36-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR