6-Aminonicotinamide 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD3623-100GEA
12170
DKK
BLDPBD3623-100G
BLDPBD3623-250MG
BLDPBD3623-25G
BLDPBD3623-5G
BLDPBD3623-10G
BLDPBD3623-1G
6-Aminonicotinamide 97%
6-Aminonicotinamide
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₆H₇N₃O Molekylvægt: 137,14 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006327 CAS-nummer: 329-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...