5-Acetyl-2,3-dichloropyridine 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD361057-100MGEA 210 DKK
BLDPBD361057-100MG
5-Acetyl-2,3-dichloropyridine 95%
5-Acetyl-2,3-dichloropyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₅Cl₂NO
Molekylvægt: 190,027 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10697703
CAS-nummer: 120800-05-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR