1-(2-Amino-4-methoxy-3-methylphenyl)ethanone 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD35400-5GEA 1070 DKK
BLDPBD35400-5G BLDPBD35400-25G BLDPBD35400-1G BLDPBD35400-250MG
1-(2-Amino-4-methoxy-3-methylphenyl)ethanone 97%
1-(2-Amino-4-methoxy-3-methylphenyl)ethanone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₀H₁₃NO₂
Molekylvægt: 179,2200 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11042290
CAS-nummer: 912347-94-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR