3-(Hydroxymethyl)isoxazole 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD35337-250MGEA 411 DKK
BLDPBD35337-250MG BLDPBD35337-100MG BLDPBD35337-1G BLDPBD35337-5G BLDPBD35337-10G
3-(Hydroxymethyl)isoxazole 98%
3-(Hydroxymethyl)isoxazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₄H₅NO₂
Molekylvægt: 99,088 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07778413
CAS-nummer: 89102-73-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR