4-Amino-2-hydroxy-3-nitropyridine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD35290-250MGEA 388 DKK
BLDPBD35290-250MG BLDPBD35290-100MG
4-Amino-2-hydroxy-3-nitropyridine 95%
4-Amino-2-hydroxy-3-nitropyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₅N₃O₃
Molekylvægt: 155,112 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09835264
CAS-nummer: 88511-57-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR