2-Methyl-4-(piperazin-1-yl)quinoline 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD35005-5GEA 5920 DKK
BLDPBD35005-5G BLDPBD35005-250MG BLDPBD35005-1G BLDPBD35005-100MG
2-Methyl-4-(piperazin-1-yl)quinoline 97%
2-Methyl-4-(piperazin-1-yl)quinoline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₄H₁₇N₃
Molekylvægt: 227,305 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01935745
CAS-nummer: 82241-22-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR