1-(4-Bromophenyl)-N-methylmethanamine 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD34670-5GEA
442
DKK
BLDPBD34670-5G
BLDPBD34670-100G
BLDPBD34670-1G
BLDPBD34670-10G
BLDPBD34670-25G
1-(4-Bromophenyl)-N-methylmethanamine 97%
1-(4-Bromophenyl)-N-methylmethanamine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₈H₁₀BrN Molekylvægt: 200,08 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04632143 CAS-nummer: 699-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...