3-Fluor-4-nitroanilin 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD3403-5GEA 132 DKK
BLDPBD3403-5G BLDPBD3403-100G BLDPBD3403-10G BLDPBD3403-25G
3-Fluor-4-nitroanilin 98%
3-Fluor-4-nitroanilin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₅FN₂O₂
Molekylvægt: 156,12 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03412239
CAS-nummer: 2369-13-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR