1,3-Dimethyl-4-piperidone 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD33717-10GEA 140 DKK
BLDPBD33717-10G BLDPBD33717-5G BLDPBD33717-100G BLDPBD33717-25G
1,3-Dimethyl-4-piperidone 97%
1,3-Dimethyl-4-piperidone

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₁₃NO
Molekylvægt: 127,1800 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00100893
CAS-nummer: 4629-80-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR