2-Chloro-6-hydroxynicotinic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD33474-100MGEA 54.3 DKK
BLDPBD33474-100MG BLDPBD33474-25G BLDPBD33474-100G BLDPBD33474-250MG BLDPBD33474-10G BLDPBD33474-1G BLDPBD33474-5G
2-Chloro-6-hydroxynicotinic acid 97%
2-Chloro-6-hydroxynicotinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₆H₄ClNO₃
Molekylvægt: 173,5500 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09757508
CAS-nummer: 38025-90-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR