1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD33128-250MGEA 117 DKK
BLDPBD33128-250MG BLDPBD33128-5G BLDPBD33128-1G
1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid 98%
1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₁₃N₃O₂
Molekylvægt: 207,23 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00664474
CAS-nummer: 303144-44-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR