7-methylchroman-4-one 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD330609-100MGEA 946 DKK
BLDPBD330609-100MG BLDPBD330609-250MG
7-methylchroman-4-one 95+%
7-methylchroman-4-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀O₂
Molekylvægt: 162,185 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00208231
CAS-nummer: 18385-69-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR