2-Fluoroisonicotinic acid 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD3276-100GEA
1420
DKK
BLDPBD3276-100G
BLDPBD3276-1G
BLDPBD3276-5G
BLDPBD3276-10G
BLDPBD3276-500G
BLDPBD3276-25G
2-Fluoroisonicotinic acid 97%
2-Fluoroisonicotinic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₆H₄FNO₂ Molekylvægt: 141,10 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02181194 CAS-nummer: 402-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...