1-Chloroisoquinoline-7-carboxaldehyde 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD32726-100MGEA 1110 DKK
BLDPBD32726-100MG BLDPBD32726-250MG
1-Chloroisoquinoline-7-carboxaldehyde 95%
1-Chloroisoquinoline-7-carboxaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₆ClNO
Molekylvægt: 191,614 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09835329
CAS-nummer: 223671-53-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR