4-Amino-3-iodobenzoic acid 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD32622-1GEA 326 DKK
BLDPBD32622-1G
4-Amino-3-iodobenzoic acid 96%
4-Amino-3-iodobenzoic acid
Formel: C₇H₆INO₂
Molekylvægt: 263,03 g/mol
MDL Number: MFCD01660550
CAS-nummer: 2122-63-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR