3-Chloro-4-ethoxyphenylboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD32310-1GEA 233 DKK
BLDPBD32310-1G BLDPBD32310-100G BLDPBD32310-25G BLDPBD32310-5G
3-Chloro-4-ethoxyphenylboronic acid 98%
3-Chloro-4-ethoxyphenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₀BClO₃
Molekylvægt: 200,4300 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03701556
CAS-nummer: 279261-81-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR