4-(Pentyloxy)phenylboronic acid 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD31972-250MGEA 46.6 DKK
BLDPBD31972-250MG BLDPBD31972-1G
4-(Pentyloxy)phenylboronic acid 97%
4-(Pentyloxy)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₇BO₃
Molekylvægt: 208,06 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04039035
CAS-nummer: 146449-90-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR