Farnesyl thiosalicylic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: FTSSalirasib

BLDPBD316909-1GEA 3710 DKK
BLDPBD316909-1G BLDPBD316909-250MG BLDPBD316909-100MG
Farnesyl thiosalicylic acid 98%
Farnesyl thiosalicylic acid

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₂₂H₃₀O₂S
Molekylvægt: 358,54 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00467723
CAS-nummer: 162520-00-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR