2,1,3-Benzothiadiazole-4-sulphonyl chloride 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD3169-1GEA 683 DKK
BLDPBD3169-1G BLDPBD3169-25G BLDPBD3169-5G
2,1,3-Benzothiadiazole-4-sulphonyl chloride 98%
2,1,3-Benzothiadiazole-4-sulphonyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₃ClN₂O₂S₂
Molekylvægt: 234,68 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00068049
CAS-nummer: 73713-79-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR