N(α)-Acetyl-L-tryptophan 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD31645-1GEA 46.6 DKK
BLDPBD31645-1G BLDPBD31645-25G BLDPBD31645-100G BLDPBD31645-500G BLDPBD31645-10G BLDPBD31645-0.5G BLDPBD31645-5G
N(α)-Acetyl-L-tryptophan 98%
N(α)-Acetyl-L-tryptophan

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₄N₂O₃
Molekylvægt: 246,3 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00065976
CAS-nummer: 1218-34-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR