3-(2-Propynylcarbamoyl)phenylboronic acid pinacol ester 95+%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD314903-1GEA 845 DKK
BLDPBD314903-1G BLDPBD314903-250MG BLDPBD314903-100MG
3-(2-Propynylcarbamoyl)phenylboronic acid pinacol ester 95+%
3-(2-Propynylcarbamoyl)phenylboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₆H₂₀BNO₃
Molekylvægt: 285,1459 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD20275029
CAS-nummer: 1357387-28-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR