5-Fluoro-2-(trifluoromethoxy)aniline 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD314548-1GEA 148 DKK
BLDPBD314548-1G BLDPBD314548-100G BLDPBD314548-0.025 BLDPBD314548-25G BLDPBD314548-5G
5-Fluoro-2-(trifluoromethoxy)aniline 98%
5-Fluoro-2-(trifluoromethoxy)aniline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₅F₄NO
Molekylvægt: 195,11 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD16652514
CAS-nummer: 123572-63-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR