4-(Difluoromethyl)benzoic acid 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD308362-5GEA 3000 DKK
BLDPBD308362-5G BLDPBD308362-1G BLDPBD308362-250MG
4-(Difluoromethyl)benzoic acid 95%
4-(Difluoromethyl)benzoic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₆F₂O₂
Molekylvægt: 172,1289 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07784270
CAS-nummer: 55805-21-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR