2,6-Dichloroisonicotinonitrile 98%
Leverandør: BLD PHARMATECH GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD3081-25GEA
1740
DKK
BLDPBD3081-25G
BLDPBD3081-250MG
BLDPBD3081-1G
BLDPBD3081-100G
BLDPBD3081-10G
BLDPBD3081-5G
2,6-Dichloroisonicotinonitrile 98%
2,6-Dichloroisonicotinonitrile
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₆H₂Cl₂N₂ Molekylvægt: 172,9995 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00173784 CAS-nummer: 32710-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...