Pentafluorophenyl-4-nitrobenzenesulphonate 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD306239-5GEA
326
DKK
BLDPBD306239-5G
BLDPBD306239-1G
BLDPBD306239-250MG
BLDPBD306239-25G
Pentafluorophenyl-4-nitrobenzenesulphonate 98%
Pentafluorophenyl-4-nitrobenzenesulphonate
Storage condition: Sealed in dry, 2-8°C
Formel:
C₁₂H₄F₅NO₅S Molekylvægt: 369,2209 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD05975120 CAS-nummer: 244633-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...