3-Amino-3-(3-bromophenyl)propionic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD30398-5GEA 396 DKK
BLDPBD30398-5G BLDPBD30398-250MG BLDPBD30398-1G BLDPBD30398-25G
3-Amino-3-(3-bromophenyl)propionic acid 97%
3-Amino-3-(3-bromophenyl)propionic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₀BrNO₂
Molekylvægt: 244,0852 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00995843
CAS-nummer: 117391-50-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR