6-Iodoindoline 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD30350-5GEA 4920 DKK
BLDPBD30350-5G BLDPBD30350-250MG BLDPBD30350-1G
6-Iodoindoline 98%
6-Iodoindoline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₈IN
Molekylvægt: 245,06 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07371641
CAS-nummer: 115666-46-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR