5-Bromobenzene-1,2,3-triol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD303179-5GEA 5480 DKK
BLDPBD303179-5G BLDPBD303179-250MG BLDPBD303179-1G BLDPBD303179-100MG
5-Bromobenzene-1,2,3-triol 95%
5-Bromobenzene-1,2,3-triol

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₅BrO₃
Molekylvægt: 205,006 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD22555992
CAS-nummer: 16492-75-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR