1,3-Benzothiazole-4-sulphonyl chloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD303141-250MGEA 2110 DKK
BLDPBD303141-250MG BLDPBD303141-100MG BLDPBD303141-1G
1,3-Benzothiazole-4-sulphonyl chloride 95%
1,3-Benzothiazole-4-sulphonyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₄ClNO₂S₂
Molekylvægt: 233,695 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD16251206
CAS-nummer: 149575-65-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR