1,1'-Azobis(N,N-dimethylformamide) 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD30023-1GEA
54.3
DKK
BLDPBD30023-1G
BLDPBD30023-10G
1,1'-Azobis(N,N-dimethylformamide) 97%
1,1'-Azobis(N,N-dimethylformamide)
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formel:
C₆H₁₂N₄O₂ Molekylvægt: 172,19 g/mol Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00008318 CAS-nummer: 10465-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...