3-(Trifluoromethyl)picolinic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD29796-25GEA 1600 DKK
BLDPBD29796-25G BLDPBD29796-1G BLDPBD29796-5G BLDPBD29796-250MG BLDPBD29796-10G
3-(Trifluoromethyl)picolinic acid 97%
3-(Trifluoromethyl)picolinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₄F₃NO₂
Molekylvægt: 191,1100 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07375386
CAS-nummer: 87407-12-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR