3-Acetyl-5-bromo-1H-indazole 96%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD293359-250MGEA 745 DKK
BLDPBD293359-250MG BLDPBD293359-1G BLDPBD293359-100MG
3-Acetyl-5-bromo-1H-indazole 96%
3-Acetyl-5-bromo-1H-indazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₇BrN₂O
Molekylvægt: 239,069 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07374362
CAS-nummer: 886363-74-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR