3-Methyl-2-(methylthio)benzo[d]thiazol-3-ium 4-methylbenzenesulphonate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD290003-1GEA 1440 DKK
BLDPBD290003-1G BLDPBD290003-100MG BLDPBD290003-250MG BLDPBD290003-5G
3-Methyl-2-(methylthio)benzo[d]thiazol-3-ium 4-methylbenzenesulphonate 95%
3-Methyl-2-(methylthio)benzo[d]thiazol-3-ium 4-methylbenzenesulphonate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₀NS₂·C₇H₇O₃S
Molekylvægt: 367,506 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00031763
CAS-nummer: 55514-14-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR