(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD288564-250MGEA
1140
DKK
BLDPBD288564-250MG
BLDPBD288564-100MG
BLDPBD288564-5G
BLDPBD288564-25G
BLDPBD288564-1G
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected 97%
(2S,4R)-4-Aminopyrrolidine-2-carboxylic acid, 4-BOC, N1-FMOC protected
Storage condition: Sealed in dry, 2-8°C
Formel:
C₂₅H₂₈N₂O₆ Molekylvægt: 452,5 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01860707 CAS-nummer: 273222-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...